Learn More
Cayman Chemical 2fluoro PalmItIc ACD mthyl 5mg
SDP
Supplier:  Cayman Chemical 245265 MG
Encompass_Preferred
A monofluorinated form of palmitic acid methyl ester; intermediate in the synthesis of 2-iodoaldehydes, monofluorinated fatty acids, and polyfluoro ketones
Catalog No. 50-293-6552