Learn More
Medchemexpress LLC 2H-1-benzopyran-2-one, 3-azido-7-hydroxy | 817638-68-9 | MFCD12924840 | 98.0% | 203.15 g/mol | C9H5N3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW54097850MG
Encompass_Preferred
3-Azido-7-hydroxycoumarin is an azide-functionalized coumarin derivative used as a fluorogenic reagent for bioorthogonal click chemistry. It combines a reactive azide with a fluorescent coumarin core to enable conjugation to alkyne-containing molecules and sensitive fluorescence detection in labeling and imaging applications.
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition.
  • Fluorescent coumarin core enables sensitive fluorescence readout.
  • Suitable for labeling biomolecules and small-molecule probes.
  • High purity supports reproducible analytical and biological assays.
  • Solid, yellow-to-brown powder with recommended storage at -20°C.
Catalog No. 50-002-84897