Learn More
Medchemexpress LLC 2H-1-benzopyran-3-carboxylic acid, 6-bromo-2-oxo- | 2199-87-3 | MFCD00047640 | 99.6% | 269.05 g/mol | C10H5BrO4 | 25 G
SDP
Supplier:  Medchemexpress LLC HY10066725G
Encompass_Preferred
UBP608 is a research-grade small molecule that acts as a potent negative allosteric modulator of N-methyl-D-aspartate receptors (NMDARs). Supplied as a white to light yellow solid with high assay purity, it is intended for laboratory studies of neurological disorders and is not for human or clinical use.
  • Potent negative allosteric modulation of NMDARs.
  • High purity (99.6% by LCMS).
  • White to light yellow solid appearance.
  • Molecular formula C10H5BrO4 and molecular weight 269.05 g/mol.
  • Available in multiple quantities, including 25 G.
  • Intended for research use only, not for human use.
Catalog No. 50-002-14207