Learn More
Medchemexpress LLC 2H-1-benzopyran, 6,7-dimethoxy-2,2-dimethyl- | 644-06-4 | 100.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1229405MG
Encompass_Preferred
Precocene II is a chromene-class insect antijuvenile hormone used in entomological and insect physiology research to study juvenile hormone antagonism and effects on development.
  • Botanical-derived chromene compound used as a research reagent.
  • Molecular formula C13H16O3 and molecular weight 220.26 g/mol.
  • High purity research grade suitable for biochemical assays and insect physiology studies.
  • Typically supplied as a 5 mg vial for small-scale experimental use.
Catalog No. 50-002-32106