Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H-1-Benzopyran-6-carbonitrile, 3,4-dihydro-2,2-dimethyl-4-(2-oxo-1-pyrrolidinyl)-3-(phosphonooxy)- | 1802655-72-6 | C16H19N2O6P | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY152847

Encompass_Preferred

Foslevcromakalim (QLS-101) is an ATP-sensitive potassium channel opener and is used as a proagent for ocular hypotensive effect.

  • ATP-sensitive potassium channel opener
  • Proagent for ocular hypotensive effect
  • High purity: 99.52%
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-75960


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.