Learn More
Medchemexpress LLC 2H-1-Benzopyran-8-carboxaldehyde, 7-hydroxy-4-methyl-2-oxo- | 14003-96-4 | 99.65% | 204.18 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY19707
Encompass_Preferred
4μ8C (IRE1 Inhibitor III) is a small-molecule inhibitor of IRE1α. It inhibits Xbp1 splicing in ER stressed cultured cells in a concentration-dependent manner. This compound plays a role in cellular and in vivo research by modulating various biological activities related to ER stress and immune responses.
  • Inhibits Xbp1 splicing in ER stressed cultured cells
  • Prevents XBP1 mRNA splicing in response to ER stress
  • Significantly inhibits IL-4, IL-5, and IL-13 production by CD4+ T cells
  • Reduces atherosclerotic lesion area in ApoE-/- mice
  • Mitigates plaque development
  • Suppresses passive cutaneous anaphylaxis in mice
  • Reverses ER stress-dependent loss of RIDD targets
Catalog No. 50-003-65654