Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H,2H,3H,3H-perfluorooctanoic acid | 914637-49-3 | 99.8% | 342.10 g/mol | C8H5F11O2 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW081085

Encompass_Preferred

2H,2H,3H,3H-perfluorooctanoic acid is an isotopically labeled perfluorooctanoic acid used as a reference standard and tracer in analytical and biochemical research. It is supplied as a white to off-white solid with high purity and a molecular weight of 342.10 g/mol.

  • Isotopically labeled with 2H and 3H.
  • High purity: 99.8% (GC).
  • Appearance: white to off-white solid.
  • Molecular weight: 342.10 g/mol.
  • Applications: analytical reference, tracer studies, assay development.

Catalog No. 50-002-09046


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.