Learn More
Medchemexpress LLC 2H,5H-Benzopyrano[4,3-b]-1,4-oxazin-9-ol, 3,4,4a,10b-tetrahydro-4-propyl-, hydrochloride | 300576-59-4 | 98.9% | 285.77 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY11000050MG
Encompass_Preferred
(+)-PD 128907 hydrochloride is a biochemical reagent used as a biological material or organic compound for life science research. It is a selective dopamine D2/D3 receptor agonist. It displaces [3H]spiperone binding from dopamine D3 receptors with high selectivity over dopamine D2 receptors. In vivo, it significantly decreases dialysate DA levels in D3 knock out mice.
  • Potent and selective dopamine D3 receptor agonist
  • Used as a biochemical reagent for life science research
  • Shows high selectivity over dopamine D2 receptors
  • Decreases dialysate DA levels in D3 knock out mice
Catalog No. 50-003-45465