Learn More
Medchemexpress LLC 2H,6H-Pyrano[3,2-b]xanthen-6-one, 3,4-dihydro-5,9-dihydroxy-7-(3-hydroxy-3-methylbutyl)-8-methoxy-2,2-dimethyl- | 26063-96-7 | 428.47 | 1 MG

Supplier: Medchemexpress LLC HYN18431MG
3-Isomangostin hydrate is a bacteriostat identified by MedChemExpress. It is an off-white to light yellow solid with a purity of 99.47%. This compound has a molecular formula of C24H28O7 and a molecular weight of 428.47. It is recommended to store the powder at -20°C for up to 3 years, and in solvent at -80°C for 6 months or -20°C for 1 month.
- Bacteriostat
- Off-white to light yellow solid appearance
- Purity of 99.47%
- Powder storage at -20°C for 3 years
- In solvent storage at -80°C for 6 months or -20°C for 1 month
- Molecular formula: C24H28O7
- Molecular weight: 428.47
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.