Learn More
Medchemexpress LLC 2H-Benzo[a]quinolizin-2-ol, 1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-, (2R,3R,11bR)- | 85081-18-1 | 98.56% | 319.44 | 200 MG
SDP
Supplier:  Medchemexpress LLC 200MGHY15793
Encompass_Preferred
NBI-98782 is a high-affinity and selective vesicular monoamine transporter 2 (VMAT2) inhibitor with a Ki of 3 nM, possessing antipsychotic activity. It is for research use only.
  • High affinity and selectivity vesicular monoamine transporter 2 (VMAT2) inhibitor
  • Ki of 3 nM
  • Exhibits antipsychotic activity
  • Solid appearance, white to off-white color
  • For research use only
Catalog No. 50-003-66217