Learn More
Medchemexpress LLC 2H-Indol-2-one, 1,3-dihydro-3-[[4-(1-methylethyl)phenyl]methylene]- | 186611-55-2 | 99.31% | 263.33 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY21291
Encompass_Preferred
SU-4313 is a potent modulator of protein tyrosine kinases (PTKs). It has the potential to modulate tyrosine kinase signal transduction to help regulate abnormal cell proliferation.
  • Potent PTKs modulator
  • Modulates tyrosine kinase signal transduction
  • Regulates abnormal cell proliferation
  • Potent against PDGFR, FLK-1, EGFR, HER2 Kinase, and IGF-1R with specific IC50 values
  • For research use only
Catalog No. 50-004-02500