Learn More
Medchemexpress LLC 2H-Indol-2-one, 1,3-dihydro-3-hydroxy-3-(2-oxo-2-phenylethyl)-1-(2-phenylethyl)- | 421578-93-0 | 99.8% | 371.43 g·mol⁻¹ | C24H21NO3 | 5 MG

Supplier: Medchemexpress LLC HY1418405MG
GLP-1R modulator C5 is a small-molecule allosteric modulator that enhances GLP-1 binding to the GLP-1 receptor via a transmembrane site. It shows in vitro activity (EC50 1.59 ± 0.53 μM) and is provided for research applications with supporting characterization and handling data.
- Allosteric enhancement of GLP-1 binding (EC50 1.59 ± 0.53 μM).
- High reported purity of 99.81%.
- Molecular formula C24H21NO3; molecular weight 371.43 g·mol⁻¹.
- Soluble in DMSO at 50 mg/mL; ultrasonic assistance recommended.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
- Available in multiple small package sizes suitable for laboratory testing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.