Learn More
Medchemexpress LLC 2H-Pyran-2,4(3H)-dione, 3-acetyl-6-methyl- | 520-45-6 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB12115G
Encompass_Preferred
Dehydroacetic acid (Biocide 470F) is a pyrone derivative with antibacterial, antifungal, and phytotoxic activities. It can be used in cosmetics, food, and beverages for preservation. Additionally, it can be utilized in the synthesis of various heterocycles and cycloaddition products.
  • Antibacterial activity
  • Antifungal activity
  • Phytotoxic activity
Catalog No. 50-004-13993