Learn More
Medchemexpress LLC 2H-tetrazole-5-acetic acid, α-amino | 138199-51-6 | MFCD00233633 | 96.5% | 143.10 g·mol⁻¹ | C3H5N5O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1008395MG
Encompass_Preferred
(RS)-(Tetrazol-5-yl)glycine is a research-grade small molecule that acts as a potent and selective N-methyl-D-aspartate (NMDA) receptor agonist. It is used in neuroscience pharmacology to probe NMDA receptor subtypes, characterize receptor pharmacology, and elicit excitatory responses in vitro and in vivo models.
  • Potent NMDA receptor agonist with documented subtype selectivity and in vivo activity.
  • High purity (96.5%) suitable for research applications.
  • Water-soluble (~5 mg/mL) with ultrasonic assistance recommended for dissolution.
  • White to off-white solid for straightforward handling and formulation.
  • Recommended storage conditions support extended stability when frozen.
Catalog No. 50-002-11388