Learn More
Medchemexpress LLC 2H-tetrazole-5-acetic acid, α-amino | 138199-51-6 | MFCD00233633 | 96.5% | 143.10 g/mol | C3H5N5O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10083950MG
Encompass_Preferred
(RS)-(Tetrazol-5-yl)glycine is a research-grade NMDA receptor agonist used in pharmacology and neuroscience to probe NMDA receptor subtype function. It exhibits subtype-selective potency in vitro and produces measurable in vivo responses in rodents. The material is supplied as a characterized solid with documented purity and solubility for experimental applications.
  • Activates NMDA receptor subtypes with distinct potency.
  • Enables pharmacological characterization of NMDA receptor function.
  • Produces in vivo responses such as seizure induction and Fos expression in rodents.
  • Water soluble for preparation of aqueous solutions (requires ultrasonication at higher concentrations).
  • Provided with certificate of analysis, datasheet, and safety documentation.
Catalog No. 50-002-11367