Learn More
Cayman Chemical 2ImIno4methylpIperIdIna 10mg
SDP
Supplier:  Cayman Chemical 8101510 MG
Encompass_Preferred
2-Imino-4-methylpiperidine is a potent, general NOS inhibitor. It exhibits IC50 values of 0.1, 1.1, and 0.2 µM for inhibition of human iNOS, eNOS, and nNOS, respectively, at an arginine concentration of 30 µM.{5579}
Catalog No. 50-289-3798