Learn More
CELLTREAT Scientific Products 2L Fernbach Flask, Vent Cap, Plain Bottom, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229860
Encompass_Preferred
CELLTREAT Scientific Products 2L Fernbach Flask. Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Conveniently pre-sterilized. 70mm opening for easy filling and dispensing. Over-sized vent area in cap for improved gas exchange. Fernbach flask shape increases surface area to volume ratio. Vent cap with plain bottom and maximum volume of 2400mL. Non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 6/case.
Catalog No. 50-202-097