Learn More
Bellco Glass, Inc. 2L Fernbach Flask Vent Cap
SDP
Supplier:  Bellco Glass, Inc. 229860000
Encompass_Preferred
Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Fernbach flask shape increases surface area to volume ratio. The baffled bottom option improves mixing and aeration.Specifications Conveniently presterilized 70mm opening for easy filling and dispensing Oversized vent area in cap for improved gas exchange Transfer Assembly available for aseptic transfer of material Size 2L Cap Type Vent Bottom Type Plain Max Volume 2200mL Case Quantity 6
Catalog No. 50-232-9713