Learn More
Teknova 2M Sorbitol. 1000mL, Sterile. 2 pack.
SDP
Supplier:  Teknova S20012PK
Encompass
Sugar is a carbohydrate that serves as a carbon source for microorganisms and can be used to test for the presence of enzymes or other proteins.
Catalog No. 50-238-7283