Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag bioMIXdrive 4 AC/DC i1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6788131EA
Encompass
Warming and maintenance-free magnetic stirrer with 4 stirring points with large surface up to 5000 mL stirring volume per point also suitable for viscous culture media.Maintenance-free 2mag-stepper-motor-drive for extremely slow and 100% jerk-free and gentle mixing of cell cultures extended speed range from 5 up to 250 rpm jerk-free even and smooth stirring also at slow speeds suitable for culture flasks with spinning propeller- and spinning ball systems.Fully encapsulated and hermetically closed stainless steel housing IP68 water- dust- and germ-proof extremely robust construction and performance easy to clean and maximum firm submersible in water baths designed for the warming-free long term operation in CO2-incubators individual measurements on request.2mag bioMIXcontrol S unit is needed sold separately.
Catalog No. 50-290-8647
Add to Cart
Request a Quote