Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag MIX 1 magnetic stirre1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6785701EA
Encompass
Ultra flat wear-free magnetic stirrer with 1 large stirring position up to 10 000 mL stirring volume also suitable for viscous media.100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerful mixing of SOLUTIONons extremely wide speed range from 100 to 2 000 rpm jerk-free stirring even at low speeds 4-step power settings for high power with regard to large and viscous stirring volumes and reduced power for the warming-free continuous operation.Maintenance-freeExtremely wide speed rangeSoftStar setting for reliable jerk-free centering of stirring barClear digital display
Catalog No. 50-290-8785