Learn More
Cayman Chemical 2MethoxymethcathInonhydro 10mg
SDP
Supplier:  Cayman Chemical 900118610 MG
Encompass_Preferred
A positional isomer of methedrone, having the methoxy group at the 2 rather than the 4 position; intended for forensic and research applications
Catalog No. 50-289-3439