Learn More
Cayman Chemical 2MethylbutyrylLcarnItInd 5mg
SDP
Supplier:  Cayman Chemical 265725 MG
Encompass_Preferred
An internal standard for the quantification of 2-methylbutyryl-L-carnitine by GC- or LC-MS
Catalog No. 50-293-3830