Learn More
Pacer Global LLC 2ml microcentrifuge tube, Non-sterile, Low retention, clear, 250 per pack
SDP
Supplier:  Pacer Global LLC MLSMTNC2/DEL
Encompass_Preferred
2ml Microcentrifuge Tube, Non-Sterile, Low retention, Natural Color, Flip Top Snap Cap.Additional info on MLS-MTNC-2: Polypropylene. Super Clear Color. Low Binding. Certified Rase/DNase free. Certified Human DNA free. Certified PCR Inhibitor Free. Certified Pyrogen Free. Leak proof snap cap. Graduation Markings. Superior strength- RCF rating of 25,000G. Temperature rating from -80° C to 121° C. Autoclavable. 200/pack.
Catalog No. NC2017156
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product