Learn More
CELLTREAT Scientific Products 2mL Pipet, Open End, Individually Wrapped, Bag, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229222B
Encompass_Preferred
"Ideal for viscous and high particulate solutionsWide, open end is ideal for slurries and gelsAll serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934; Accuracy is ±2%Bi-directional (ascending & descending) graduations are printed in bold easy-to-read black inkPremium polymer fiber barrier plug outperforms cotton plugsColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controller"
Catalog No. NC3433446