Learn More
USA Scientific Inc 2.0 mL conical screw cap tube, natural
Supplier:  USA Scientific Inc 14208700
Encompass_Preferred
2.0 mL conical base screw cap microcentrifuge tube, graduated, natural with natural cap, 500/pack
Catalog No. NC9676635