Learn More
Elemental Scientific Inc 2ML SAMPLE LOOP 3.2X1.6MM
SDP
Supplier:  Elemental Scientific Inc SC031520
Encompass_Preferred
Recommended for use with 1.0 mm i.d. (gray) probes and applications requiring larger sample volumes or high flow rates. 3.2 mm o.d., 1/4-28 fittingsApplications- soilFAST, oilFAST, and large volume FAST
Catalog No. NC2595862