Learn More
Sarstedt Inc 2.0ml SC Micro Tube PCR-PT. 1000/Pack
Supplier: Sarstedt Inc 72.694.406
Screw Cap Micro Tube, 2 ml, PP, with smear edge, with knurls, with assembled cap, with writing space and graduation, certified PCR Performance Tested (free of DNase/RNase, DNA and PCR inhibitors), 100 pcs./bag