Learn More
Bellco Glass, Inc. 2ml Serological Pipette, Sterile, 20 per bulk bag, 1000/pk, 3000/cs
SDP
Supplier:  Bellco Glass, Inc. 451400250
Encompass_Preferred
2ml Serological Pipette with 0.6ml negative graduation, 1/100 ml graduations, Sterile, 20 per bulk bag, 1000/pk, 3000/cs. Use for liquid transfer. bulk packaging serologcials offer the same high quality serological pipettes, but in a more environment friendly way.
Catalog No. 50-232-9676