Learn More
IBI SCIENTIFIC 3P 2ML X 10 COMB
Supplier:  IBI SCIENTIFIC 3P IB52070
Encompass_Preferred
IBI’s MP-1015 2.0mm x 10 tooth comb is compatible with the MP-1015 Horizontal Electrophoresis System (IB53000.) IBI combs are machined from Delrin to provide years of service.•Well Width: 6.3mm;•Sample Volume per mm Gel Thickness: 12.6ul
Catalog No. NC0838528