Learn More
Chemglass Life Sciences Stopcock, Double Oblique Bore, Glass Plug, 2mm Bore, 14.5/50 Plug Size, 8mm OD Sidearms
SDP
Supplier:  Chemglass Life Sciences CG41401
Encompass_Preferred
Stopcock, Double Oblique Bore, Glass Plug, 2mm Bore, 14.5/50 Plug Size, 8mm OD Sidearms. Stopcock has a solid glass plug having parallel diagonal bores. Both sizes are supplied complete with metal retaining clip CG-423.
Catalog No. CG41401