Learn More
Teknova 2mM Sodium Citrate pH 6.5, 1000mL, Endotoxin/RNase/DNase/Protease Tested, Sterile. 8 pack.
SDP
Supplier:  Teknova S81658PK
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-7665