Learn More
Medchemexpress LLC HY-102081 2mg Medchemexpress, 2OH-BNPP1 CAS:833481-73-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1020812MG
Encompass_Preferred
Medchemexpress, HY-102081 2mg 2OH-BNPP1 CAS:833481-73-5 2OH-BNPP1 is an inhibitor of BUB1 kinase, a Ser/Thr kinase, used for the treatment of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7979