Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2OH-BNPP1 | 833481-73-5 | MFCD28120704 | 99.7% | 297.35 g/mol | C16H19N5O | 10 MG
SDP

Supplier:  Medchemexpress LLC HY10208110MG

Encompass_Preferred

2OH-BNPP1 is a small-molecule inhibitor of the BUB1 serine/threonine kinase supplied for research use. It is provided as a high-purity solid suitable for in vitro and in vivo studies, with documented solubility profiles and recommended storage conditions to preserve stability.

  • High purity (99.65%).
  • Molecular formula C16H19N5O; molecular weight 297.35 g/mol.
  • CAS number 833481-73-5.
  • White to off-white solid appearance.
  • Soluble in DMSO (100 mg/mL) and can be formulated for in vivo use.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (2 years).
  • Available in multiple pack sizes, including a 10 mg pack.

Catalog No. 50-002-48479


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.