Learn More
Cayman Chemical 2Omethyl PAF C18hydrAT 10mg
SDP
Supplier:  Cayman Chemical 6091210 MG
Encompass_Preferred
A synthetic PAF analog which contains an ether-linked methyl group at the sn-2 position; decreases plasma membrane fluidity, inhibits tumor cell invasiveness, and causes the release of large quantities of NO by a pathway involving activation of NOS
Catalog No. 50-289-4985