Learn More
MSC Leviton 2341 480 VAC 20A NEMA L8-20P Industrial Twist Lock Plug 2 Poles, 3 Wires, 1 Phase, Self-Grounding, 0.385 to 0.78" Cord Diam, Black/White
Supplier:  MSC 74364704
Encompass
Leviton 2341 480 VAC 20A NEMA L8-20P Industrial Twist Lock Plug 2 Poles, 3 Wires, 1 Phase, Self-Grounding, 0.385 to 0.78" Cord Diam, Black/White
Catalog No. 50-129-7746