Learn More
Medchemexpress LLC (2R)-2-hydroxy-2-methyl-4-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)butanamide | 1147883-03-1 | MFCD00824427 | 99.3% | 265.30 g/mol | C14H19NO4 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12599950MG
Encompass_Preferred
EPI-589 is a redox-active quinone derivative investigated as a neuroprotective small molecule. It acts as an oxidoreductase inhibitor and free radical scavenger, and has demonstrated blood-brain barrier permeability and oral bioavailability in preclinical and early clinical studies. It is used in research on neurodegenerative diseases to study oxidative stress and mitochondrial dysfunction.
  • Redox-active oxidoreductase inhibitor and free radical scavenger.
  • Blood-brain barrier permeable and orally bioavailable.
  • Demonstrated neuroprotective activity in preclinical models.
  • High purity suitable for research applications.
  • Soluble in DMSO; hygroscopic-use freshly opened solvent.
  • Stable as a powder when stored at -20°C or refrigerated per supplier guidance.
Catalog No. 50-002-38071