Learn More
Medchemexpress LLC (2R)-3-(1H-indol-3-yl)-2-[[5-[2-(4-methylphenyl)ethynyl]thiophen-2-yl]sulfonylamino]propanoic acid | 203640-27-1 | MFCD22200275 | 99.8% | 464.56 | C24H20N2O4S2 | 50 MG

Supplier: Medchemexpress LLC HY10699250MG
S 3304 is a research-grade small-molecule inhibitor of matrix metalloproteinases, primarily used in biochemical and cell-based studies targeting MMP activity. The compound is provided as a light yellow solid and is intended for laboratory research use only, with recommended cold-storage conditions to preserve stability.
- High chemical purity: 99.8%.
- Molecular weight: 464.56 g·mol⁻¹.
- Chemical formula: C24H20N2O4S2.
- Appearance: light yellow to yellow solid powder.
- Recommended storage: powder -20°C for up to 3 years; in solvent store at -80°C for long term.
- Available in milligram-scale packaging for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.