Learn More
Medchemexpress LLC (2R)-3-(((2-(2-azidoacetamido)ethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl distearate | 2839508-98-2 | 96.4% | 831.11 | C43H83N4O9P | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW44099910MG
Encompass_Preferred
DSPE-N3 is an azide-functionalized phospholipid (C43H83N4O9P; CAS 2839508-98-2) intended for research use. It provides an azide handle for covalent conjugation via copper-catalyzed or strain-promoted azide-alkyne cycloaddition, enabling facile functionalization of liposomes, nanoparticles, and surfaces.
Catalog No. 50-002-81758