Learn More
Strem, An Ascensus Company CAS# 147702-13-4. 500mg. (2R)-(+)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol, min. 98% (R)-VANOL. MFCD08459338
SDP
Supplier:  Strem, An Ascensus Company 081700500MG
Encompass
CAS# 147702-13-4. 500mg. (2R)-(+)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol, min. 98% (R)-VANOL. MFCD08459338. Molecular Weight: 438.52. Molecular Formula: C32H22O2. Color/form: white to pale yellow pwdr. Strem# 08-1700. http://www.strem.com/catalog/v/08-1700/
Catalog No. 50-901-09216