Learn More
Medchemexpress LLC (2R,3R)-butane-2,3-diol | 24347-58-8 | MFCD00064267 | 97.0% | 90.12 g/mol | C4H10O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01595410MM/1ML
Encompass_Preferred
(2R,3R)-butane-2,3-diol is the (R,R) enantiomer of 2,3-butanediol supplied as an analytical standard and as a ready-to-use solution for research and analytical applications. It is a chiral diol metabolite (CAS 24347-58-8) with molecular weight 90.12 g/mol and is offered at high purity for chromatographic and mass spectrometric method development.
  • Chiral analytical standard suitable for enantiomer-specific analysis.
  • High purity (97%) for reliable analytical results.
  • Available as a ready-to-use solution (example: 10 mM, 1 mL) for quick workflows.
  • Compatible with GC, LC, and mass spectrometry applications.
  • Stable liquid form, easy to aliquot and handle.
Catalog No. 50-003-01001