Learn More
Medchemexpress LLC (2R,3R)-butane-2,3-diol | 24347-58-8 | MFCD00064267 | 99.2% | 90.12 g/mol | C4H10O2 | 1 G

Supplier: Medchemexpress LLC HYW0159541G
(2R,3R)-Butane-2,3-diol is the chiral (2R,3R) stereoisomer of butane-2,3-diol supplied as a high-purity reference and biological probe for research into bacterial calcium signaling and microbiota-host interactions.
- High purity (99.17%).
- Molecular formula C4H10O2; molecular weight 90.12 g/mol.
- Liquid; colorless to light yellow; density 0.992 g/cm3.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
- Storage: pure form -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).
- Acts as a La3+-sensitive calcium channel agonist in bacteria; reported EC50 ≈ 25 mM.
- Available as a neat liquid in laboratory pack sizes from grams to bulk quantities.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.