Learn More
Medchemexpress LLC (2R,3R)-Firazorexton | 2692692-00-3 | 99.9% | 470.51 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY137440B1MG
Encompass_Preferred
(2R,3R)-Firazorexton, also known as (2R,3R)-TAK-994 free base, is an isomer of Firazorexton. This orally active and brain-penetrating compound acts as an orexin type 2 receptor (OX2R) agonist, showing potential in improving narcolepsy-like symptoms. It is provided as a white to off-white solid and is intended for research use only.
  • Orally active and brain-penetrating
  • Orexin type 2 receptor (OX2R) agonist
  • Potential to improve narcolepsy-like symptoms
  • Suitable for research in neurological disorders
  • Provided as a white to off-white solid
Catalog No. 50-003-48892