Learn More
Medchemexpress LLC (2R,3R)-Firazorexton | 2692692-00-3 | 99.9% | C22H25F3N2O4S | 10MG
SDP
Supplier:  Medchemexpress LLC HY137440B10MG
Encompass_Preferred
(2R,3R)-Firazorexton is an orally active, brain-penetrant orexin type 2 receptor (OX2R) agonist used in preclinical research to probe wakefulness mechanisms and narcolepsy-like behaviors. It is supplied in high-purity solid form and as a 10 mM DMSO solution for biochemical and in vivo studies.
  • High purity suitable for biochemical and pharmacological assays.
  • Orally active OX2R agonist for wakefulness research.
  • Brain-penetrant in preclinical models.
  • Available as solid and 10 mM solution in DMSO.
  • Supplied with purity and batch documentation.
  • Appropriate for in vivo and in vitro pharmacology studies.
Catalog No. 50-002-59948