Learn More
Medchemexpress LLC (2R,3R)-GSK973 | 00-00-0 | >98.0% | C23H23FN2O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY138563A10MG
Encompass_Preferred
(2R,3R)-GSK973 is the (2R,3R) stereoisomer of a BD2-selective BET bromodomain inhibitor used as a research tool in epigenetics. It is provided as a high-purity small-molecule solid for biochemical and cellular studies of BD2 domains.
Catalog No. 50-002-60583