Learn More
Medchemexpress LLC (2R,3R)-GSK973 | 410.44 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138563A
Encompass_Preferred
(2R,3R)-GSK973 is an isomer of GSK973, a highly selective and orally bioavailable inhibitor targeting the BD2s (second bromodomains) of the BET family. This compound functions within the epigenetic reader domain pathway, making it suitable for various research applications.
  • Selective inhibitor of BD2s of the BET family
  • Functions within the epigenetic reader domain pathway
  • Orally bioavailable
  • Suitable for research applications
Catalog No. 50-003-49143