Learn More
Medchemexpress LLC (2R,3S)-3-Phenylisoserine hydrochloride | 132201-32-2 | 25 G

Supplier: Medchemexpress LLC 25GHYN2378
(2R,3S)-3-Phenylisoserine hydrochloride is a serine derivative primarily for research use. As an amino acid derivative, it can be used as an ergogenic supplement, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress-related tasks, and preventing exercise-induced muscle damage.
- Serine derivative for research use
- Can influence anabolic hormone secretion
- Supplies fuel during exercise
- Improves mental performance
- Helps prevent exercise-induced muscle damage
- Appears as a white to off-white solid
- Melting point 222-224 °C
- Store at 4 °C, sealed and away from moisture
- In solvent, store at -80 °C for 6 months or -20 °C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.