Learn More
Medchemexpress LLC (2R,3S)-Azelaprag | 2049979-31-7 | 99.1% | 523.61 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1677125MG
Encompass_Preferred
(2R,3S)-Azelaprag is a precursor of an apelin receptor agonist with an EC50 of 12 nM. It is an isomer of Azelaprag and is intended for research use only.
  • Appearance: Solid
  • Color: White to off-white
  • Apelin receptor agonist
  • Isomer of Azelaprag
Catalog No. 50-003-91143