Learn More
Medchemexpress LLC (2R,3S)-Azelaprag | 2049979-31-7 | 99.1% | 523.61 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16771250MG
Encompass_Preferred
(2R,3S)-Azelaprag is a precursor of an apelin receptor agonist (EC50=12 nM) and an isomer of Azelaprag. It is for research use only.
  • Apelin receptor agonist (ec50=12 nM)
Catalog No. 50-003-90573