Learn More
Medchemexpress LLC (2R,3S)-Azelaprag | 2049979-31-7 | 99.10% | 523.61 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16771210MM/1ML
Encompass_Preferred
(2R,3S)-Azelaprag is a precursor of an apelin receptor agonist, provided as a 10 mM solution in 1 mL of DMSO. This compound, also known as (2R,3S)-AMG 986, is an isomer of Azelaprag and is intended for research use only.
  • Acts as an apelin receptor agonist with an EC50 of 12 nM.
Catalog No. 50-003-90654